Dibromomethane 99%

Supplier: Thermo Fisher Scientific
Warning

112831000 112835000 112830010 112830025
ACRO112831000EA 51.7 EUR
ACRO112831000 ACRO112835000 ACRO112830010 ACRO112830025
Dibromomethane 99%
Dibromomethane
Formula: Br₂CH₂
Boiling Pt: 96…98 °C (760 mmHg)
Melting Pt: –52 °C
Density: 2,47 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000168
CAS Number: 74-95-3
EINECS: 200-824-2
UN: 2664
ADR: 6.1,III
Merck Index: 12,06138

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5400 to 1.5420 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR