1,2-Diiodoethane 99%

Supplier: Thermo Fisher Scientific
Warning

115140250 115141000
ACRO115140250EA 59.4 EUR
ACRO115140250 ACRO115141000
1,2-Diiodoethane 99%
1,2-Diiodoethane
Formula: C₂H₄I₂
MW: 281.86 g/mol
Boiling Pt: 206
Melting Pt: 80…83 °C
Density: 2.13 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00001093
CAS Number: 624-73-7
EINECS: 210-859-5

Order Now

Specification Test Results

Appearance Brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 80 to 83 °C
GC ≥98.5 %
Clarity of solution Passes test
Substance darkened by H2SO4 Passes test
Chloride (Cl) ≤20 ppm
Nitrate (NO3) ≤20 ppm
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤200 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤20 ppm
Insoluble matter ≤0.01 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR