2,3-Dimethoxybenzoic acid 99%

Supplier: Thermo Fisher Scientific
Warning

115420050 115420250 115421000
ACRO115420050EA 23.4 EUR
ACRO115420050 ACRO115420250 ACRO115421000
2,3-Dimethoxybenzoic acid 99%
2,3-Dimethoxybenzoic acid
Formula: C₉H₁₀O₄
MW: 182.18 g/mol
Boiling Pt: 314
Melting Pt: 121…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00002432
CAS Number: 1521-38-6
EINECS: 216-188-4

Order Now

Specification Test Results

Appearance Light beige Crystalline powder
Infrared spectrum Conforms
Melting point 121 to 124 °C
HPLC ≥98.5 %
Titration with NaOH ≥98.5 %
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Sulfur compounds (as S) ≤0.005 %
Water soluble titr. acid ≤0.0003 meq/g

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR