4-(Dimethylamino)antipyrine 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2,3-Dimethyl-4-(dimethylamino)-1-phenyl-3-pyrazolin-5-one, 4-(Dimethylamino)phenazone

115740050 115741000 115740010 115740250
ACRO115740050EA 27 EUR
ACRO115740050 ACRO115741000 ACRO115740010 ACRO115740250
4-(Dimethylamino)antipyrine 97%
4-(Dimethylamino)antipyrine
Formula: C₁₃H₁₇N₃O
MW: 231.3 g/mol
Melting Pt: 105…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003142
CAS Number: 58-15-1
EINECS: 200-365-8
UN: 2811
ADR: 6.1 III
Merck Index: 12,00495

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
Melting point 105 to 110 °C
Titration with HClO4 ≥96.0 %
Specific optical rotation -15° to -18° (20 °C, 589 nm) (c=10, EtOH)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR