4-(Dimethylamino)benzoic acid 98%

Supplier: Thermo Fisher Scientific
Warning

115760250 115761000 115765000
ACRO115760250EA 55.5 EUR
ACRO115760250 ACRO115761000 ACRO115765000
4-(Dimethylamino)benzoic acid 98%
4-(Dimethylamino)benzoic acid
Formula: C₉H₁₁NO₂
Boiling Pt: 297 °C (1013 hPa)
Melting Pt: 238…242 °C
Density: 1,29 g/cm³ (20 °C)
Flash Pt: 170 °C
Storage Temperature: Ambient
MDL Number: MFCD00002537
CAS Number: 619-84-1
EINECS: 210-615-8
Merck Index: 13,03259

Order Now

Specification Test Results

Appearance White to beige to grey-green Crystalline powder
Infrared spectrum Conforms
Melting point 238 to 242 °C
Water ≤0.5 % (K.F.)
Titration with NaOH ≥97.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR