N-Ethylaniline 98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: N-Ethyl-N-phenylamine

118040025 118041000 118045000
ACRO118040025EA 257 EUR
ACRO118040025 ACRO118041000 ACRO118045000
N-Ethylaniline 98%
N-Ethylaniline
Formula: C₈H₁₁N
Boiling Pt: 204,7 °C (1013 hPa)
Melting Pt: –63 °C
Density: 0,963 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00009025
CAS Number: 103-69-5
EINECS: 203-135-5
UN: 2272
ADR: 6.1,III
Merck Index: 13,03800

Order Now

Specification Test Results

Appearance Clear light yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5530 to 1.5560 (20°C, 589 nm)
GC ≥97.5 %
Titration with HClO4 ≥99.0 %
Chloride (Cl) ≤0.002 %
Phosphate (PO4) ≤0.001 %
Sulfate (SO4) ≤0.003 %
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤0.001 %
Magnesium (Mg) ≤0.002 %
Heavy metals (ICP-OES) ≤0.001 %
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR