Ethyl carbazate 97%

Supplier: Thermo Fisher Scientific
Warning

118200250 118201000 118205000
ACRO118200250EA 48.5 EUR
ACRO118200250 ACRO118201000 ACRO118205000
Ethyl carbazate 97%
Ethyl carbazate
Formula: C₃H₈N₂O₂
Boiling Pt: 108…110 °C (22 mmHg)
Melting Pt: 44…48 °C
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00007595
CAS Number: 4114-31-2
EINECS: 223-903-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to almost white (May have a pink hue) Crystals and/or chunks or fused solid
Infrared spectrum Conforms
Melting point 42 to 48 °C
GC ≥96.0 %
Chloride (Cl) ≤20 ppm
Sulfate (SO4) ≤50 ppm
Iron (Fe) ≤20 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR