2-Fluorobenzoic acid 99%

Supplier: Thermo Fisher Scientific
Warning

119360250 119361000
ACRO119360250EA 30.7 EUR
ACRO119360250 ACRO119361000
2-Fluorobenzoic acid 99%
2-Fluorobenzoic acid
Formula: C₇H₅FO₂
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 123…125 °C
Density: 1,46 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002405
CAS Number: 445-29-4
EINECS: 207-158-1

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or needles
Infrared spectrum Conforms
Melting point 122 to 125 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR