α-Bromo-3-fluorotoulene 95%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: α-Bromo-3-fluorotoluene, α-Bromo-m-fluorotoulene, alpha-Bromo-m-fluorotoulene, 1-(Bromomethyl)-3-fluorobenzene, m-Fluorobenzyl bromide, 3-Fluorobenzyl bromide

119400250 119400050
ACRO119400250EA 121 EUR
ACRO119400250 ACRO119400050
α-Bromo-3-fluorotoulene 95%
α-Bromo-3-fluorotoulene
Formula: C₇H₆BrF
Boiling Pt: 88 °C (20 mmHg)
Density: 1,541 g/cm³
Flash Pt: 61 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000340
CAS Number: 456-41-7
EINECS: 207-263-2
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5450 to 1.5480 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR