L(+)-Glutamine 99%

Supplier: Thermo Fisher Scientific

Synonyms: Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q

119951000 119955000 119950025
ACRO119951000EA 103 EUR
ACRO119951000 ACRO119955000 ACRO119950025
L(+)-Glutamine 99%
L(+)-Glutamine
Formula: C₅H₁₀N₂O₃
Boiling Pt: 355 °C (1013 hPa)
Melting Pt: 185 °C (dec.)
Density: 1,5326 g/cm³ (130 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008044
CAS Number: 56-85-9
EINECS: 200-292-1
Merck Index: 13,04484
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb7a812ec5d2c73a797fc464926df9eef12c5c9a&vid=15d7d87daff0fec33a974a578415e157a3278049&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.3 % (105°C, 3 h)
Sulfated ash ≤0.3 %
Specific optical rotation +31.5° to +33.0° (20°C, 589 nm) (c=10, 2N HCl)
Titration with HClO4 98.5 to 101.5 %
Iron (Fe) ≤30 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR