Guaiacol 99+%

Supplier: Thermo Fisher Scientific
Warning

120190010 120192500 120190050
ACRO120190010EA 111 EUR
ACRO120190010 ACRO120192500 ACRO120190050
Guaiacol 99+%
Guaiacol
Formula: C₇H₈O₂
Boiling Pt: 205 °C (760 mmHg)
Melting Pt: 27…29 °C
Density: 1,12 g/cm³
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00002185
CAS Number: 90-05-1
EINECS: 201-964-7
ADR: 6.1,III
Merck Index: 13,04568

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid or crystalline mass
Infrared spectrum Conforms
Refractive index 1.5390 to 1.5450 (20°C, 589 nm)
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR