Imidazole 99%

Supplier: Thermo Fisher Scientific
Danger

122020020 122020050 122021000 122025000 122020100
ACRO122020020EA 547 EUR
ACRO122020020 ACRO122020050 ACRO122021000 ACRO122025000 ACRO122020100
Imidazole 99%
Imidazole
Formula: C₃H₄N₂
Boiling Pt: 255…256 °C (760 mmHg)
Melting Pt: 86…90 °C
Density: 1,18 g/cm³ (20 °C)
Flash Pt: 145 °C
Storage Temperature: Ambient
MDL Number: MFCD00005183
CAS Number: 288-32-4
EINECS: 206-019-2
UN: 3263
ADR: 8,III
Merck Index: 13,04935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48eba49e61201d6fffa2bef0c54633a20660c7ef&vid=e7f154e4976b52732539e49bbff60454d5eb82ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to light yellow Flakes
Infrared spectrum Conforms
Water ≤0.2 % (K.F.)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR