Iodoacetic acid 97%

Supplier: Thermo Fisher Scientific
Danger

122280250 122280500 122281000
ACRO122280250EA 91 EUR
ACRO122280250 ACRO122280500 ACRO122281000
Iodoacetic acid 97%
Iodoacetic acid
Formula: C₂H₃IO₂
Boiling Pt: 262 °C (1013 hPa)
Melting Pt: 79…83 °C
Density: 2,2694 g/cm³ (85 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002685
CAS Number: 64-69-7
EINECS: 200-590-1
UN: 2923
ADR: 8,I
Merck Index: 13,05047

Order Now

Specification Test Results

Appearance Yellow to orange-brown Powder or crystals or flakes
Infrared spectrum Conforms
Melting point 76 to 83 °C
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR