Isonicotinic acid 99%

Supplier: Thermo Fisher Scientific
Warning

122591000 122595000 122590025
ACRO122591000EA 49.2 EUR
ACRO122591000 ACRO122595000 ACRO122590025
Isonicotinic acid 99%
Isonicotinic acid
Formula: C₆H₅NO₂
Boiling Pt: 260 °C (15 mmHg)
Melting Pt: 310…315 °C
Storage Temperature: Ambient
MDL Number: MFCD00006429
CAS Number: 55-22-1
EINECS: 200-228-2
Merck Index: 13,05206

Order Now

Specification Test Results

Appearance White to cream Powder
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %
HPLC ≥97.5 %
Specific optical rotation -190° to -204° (20°C, 589 nm) (c=2,acetone)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR