Isophthaloyl dichloride 98%

Supplier: Thermo Fisher Scientific
Danger

122661000 122665000
ACRO122661000EA 34.7 EUR
ACRO122661000 ACRO122665000
Isophthaloyl dichloride 98%
Isophthaloyl dichloride
Formula: C₈H₄Cl₂O₂
Boiling Pt: 276 °C (760 mmHg)
Melting Pt: 43…45 °C
Density: 1,388 g/cm³ (17 °C)
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00000678
CAS Number: 99-63-8
EINECS: 202-774-7
UN: 2928
ADR: 6.1,I

Order Now

Specification Test Results

Appearance White Crystalline mass
Infrared spectrum Conforms
Melting point 43 to 45 °C
Titration RCOCl ≥97.5 %
NMR Consistent with structure
Specific optical rotation +30° to +33° (20 °C, 589 nm) (C= neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR