Ammonium chloride 99+%, pure

Supplier: Thermo Fisher Scientific
Warning

123340010 123340250 123340050 123340251
ACRO123340010EA 63.5 EUR
ACRO123340010 ACRO123340250 ACRO123340050 ACRO123340251
Ammonium chloride 99+%, pure
Ammonium chloride
Formula: NH₄Cl
Melting Pt: 340 °C (subl.)
Density: 1,5256 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
EINECS: 235-186-4
Merck Index: 13,00510
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=389479ce9f26d046837e12f5c47dd4bc7d47ec2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder and/or chunks
Titration Argentometric ≥99.0 %
Water ≤0.1 % (K.F.)
Sulfated ash ≤0.3 %
Iron (Fe) ≤10 ppm
Insoluble matter ≤0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR