(4-Chlorophenyl)acetyl chloride 96%

Supplier: Thermo Fisher Scientific
Danger

123450050 123450250 123451000
ACRO123450050EA 33.5 EUR
ACRO123450050 ACRO123450250 ACRO123451000
(4-Chlorophenyl)acetyl chloride 96%
(4-Chlorophenyl)acetyl chloride
Formula: (C₆H₄Cl)CH₂COCl
Boiling Pt: 85 °C (1 mmHg)
Density: 1,292 g/cm³
Flash Pt: >110 °C
MDL Number: MFCD00037111
CAS Number: 25026-34-0
EINECS: 246-571-1
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5490 to 1.5570 (20°C, 589 nm)
GC ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR