Sodium carbonate, anhydrous 99.5%, extra pure

Supplier: Thermo Fisher Scientific
Warning

123670010 123670025 123670100
ACRO123670010EA 51.7 EUR
ACRO123670010 ACRO123670025 ACRO123670100
Sodium carbonate, anhydrous 99.5%, extra pure
Sodium carbonate
Formula: Na₂CO₃
Boiling Pt: 1600 °C (760 mmHg)
Melting Pt: 851 °C
Density: 2,53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Granular powder
Loss on drying ≤1 % (300°C, 2 hrs)
Titration with HCl 99.5 to 100.5 % (On dried substance)
Chloride (Cl) ≤125 ppm
Sulfate (SO4) ≤250 ppm
Arsenic (As) ≤5 ppm
Iron (Fe) ≤50 ppm
Heavy metals (as Pb) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR