6-Methoxy-1-tetralone 99%

Supplier: Thermo Fisher Scientific

Synonyms: 6-Methoxy-1,2,3,4-tetrahydro-1-naphthalenone, 6-Methoxy-3,4-dihydronaphthalen-1(2H)-one

126120250 126121000 126125000
ACRO126120250EA 47 EUR
ACRO126120250 ACRO126121000 ACRO126125000
6-Methoxy-1-tetralone 99%
6-Methoxy-1-tetralone
Formula: C₁₁H₁₂O₂
Boiling Pt: 171 °C (11 mmHg)
Melting Pt: 77…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00001695
CAS Number: 1078-19-9
EINECS: 214-078-0

Order Now

Specification Test Results

Appearance Yellow to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 77 to 79 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR