4'-Methylacetophenone 95%

Supplier: Thermo Fisher Scientific
Warning

126180010 126180050 126182500
ACRO126180010EA 98.2 EUR
ACRO126180010 ACRO126180050 ACRO126182500
4'-Methylacetophenone 95%
4'-Methylacetophenone
Formula: C₉H₁₀O
Boiling Pt: 226 °C (1013 hPa)
Melting Pt: –19 °C
Density: 1,005 g/cm³
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00008751
CAS Number: 122-00-9
EINECS: 204-514-8

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5300 to 1.5350 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR