2'-Nitroacetophenone 95%

Supplier: Thermo Fisher Scientific
Warning

128320025 128320200
ACRO128320025EA 31.8 EUR
ACRO128320025 ACRO128320200
2'-Nitroacetophenone 95%
2'-Nitroacetophenone
Formula: C₈H₇NO₃
Boiling Pt: 159 °C (16 mmHg)
Melting Pt: 28…30 °C
Density: 1,24 g/cm³ (20 °C)
Flash Pt: >112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007144
CAS Number: 577-59-3
EINECS: 209-414-8

Order Now

Specification Test Results

Appearance Orange to brown Low melting solid
Infrared spectrum Conforms
GC ≥88 %
Stabilizer 0.05 % BHT


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR