1,1,3,3-Tetramethylbutylamine 95%

Supplier: Thermo Fisher Scientific
Danger

129500050 129505000
ACRO129500050EA 25.3 EUR
ACRO129500050 ACRO129505000
1,1,3,3-Tetramethylbutylamine 95%
1,1,3,3-Tetramethylbutylamine
Formula: C₈H₁₉N
Boiling Pt: 137 °C (1013 hPa)
Melting Pt: –67 °C
Density: 0,805 g/cm³
Flash Pt: 32 °C
MDL Number: MFCD00008053
CAS Number: 107-45-9
EINECS: 203-491-1
UN: 2734

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4235 to 1.4255 (20°C, 589 nm)
Water ≤0.3 %
GC ≥94 %
Color scale ≤25 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR