Oleyl amine ∼ 80-90% C18

Supplier: Thermo Fisher Scientific
Danger

Synonyms: cis-9-Octadecenylamine, (9Z)-9-Octadecen-1-amine, (Z)-Octadec-9-en-1-amine

129540010 129540050 129541000 129540051
ACRO129540010EA 273 EUR
ACRO129540010 ACRO129540050 ACRO129541000 ACRO129540051
Oleyl amine ∼ 80-90% C18
Oleyl amine
Formula: C₁₈H₃₇N
MW: 267.5 g/mol
Boiling Pt: 364
Melting Pt: 8…18 °C
Density: 0.81 g/cm³
Flash Pt: 154 °C
Storage Temperature: Ambient
MDL Number: MFCD00066507
CAS Number: 112-90-3
EINECS: 204-015-5
UN: 2735
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4585 to 1.4625 (20°C, 589 nm)
Titration with HClO4 ≥96.0 % (Primary amine)
Additional info dielectric strength: ≥35 kV
Viscosity 0.7 to 0.9 cSt (at 25°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR