Oxamide 98%

Supplier: Thermo Fisher Scientific
Warning

129651000 129655000
ACRO129651000EA 41.9 EUR
ACRO129651000 ACRO129655000
Oxamide 98%
Oxamide
Formula: C₂H₄N₂O₂
MW: 88.07 g/mol
Melting Pt: 491 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008007
CAS Number: 471-46-5
EINECS: 207-442-5

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration excess NaOH ≥97.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR