Trichloromethanesulphenyl chloride 97%

Supplier: Thermo Fisher Scientific
Danger

130030050 130030250
ACRO130030050EA 60 EUR
ACRO130030050 ACRO130030250
Trichloromethanesulphenyl chloride 97%
Trichloromethanesulphenyl chloride
Formula: CCl₄S
Boiling Pt: 146…148 °C (760 mmHg)
Density: 1,7 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00037054
CAS Number: 594-42-3
EINECS: 209-840-4
UN: 1670
ADR: 6.1,I

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5370 to 1.5410 (20°C, 589 nm)
Assay ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR