3-Phenylpropylamine 98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 3-Phenyl-1-propylamine

130880250
ACRO130880250EA 64.8 EUR
ACRO130880250
3-Phenylpropylamine 98%
3-Phenylpropylamine
Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 221
Density: 0.95 g/cm³
Flash Pt: 90 °C
MDL Number: MFCD00008224
CAS Number: 2038-57-5
EINECS: 218-012-1
UN: 2735

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5240 to 1.5260 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥97.5 %
Peroxides ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR