Phloroglucinol dihydrate 99%

Supplier: Thermo Fisher Scientific
Warning

131040250 131041000 131045000
ACRO131040250EA 39.4 EUR
ACRO131040250 ACRO131041000 ACRO131045000
Phloroglucinol dihydrate 99%
Phloroglucinol dihydrate
Formula: C₆H₆O₃·2H₂O
Melting Pt: 218…221 °C
Storage Temperature: Ambient
MDL Number: MFCD00149090
CAS Number: 6099-90-7
EINECS: 203-611-2
Merck Index: 13,07413

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR