Phthaloyl dichloride 90% remainder mainly phthalic acid

Supplier: Thermo Fisher Scientific
Danger

131110050
ACRO131110050EA 28.5 EUR
ACRO131110050
Phthaloyl dichloride 90% remainder mainly phthalic acid
Phthaloyl dichloride
Formula: C₈H₄Cl₂O₂
Boiling Pt: 269…271 °C (760 mmHg)
Melting Pt: 12 °C
Density: 1,4 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000666
CAS Number: 88-95-9
EINECS: 201-869-0
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5674 to 1.5694 (20°C, 589 nm)
Titration RCOCl ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR