1,3-Propanediol 98%

Supplier: Thermo Fisher Scientific

Synonyms: 1,3-Propylene glycol, 1,3-Dihydroxy propane

131400010 131402500 131400025 131400100
ACRO131400010EA 288 EUR
ACRO131400010 ACRO131402500 ACRO131400025 ACRO131400100
1,3-Propanediol 98%
1,3-Propanediol
Formula: HO(CH₂)₃OH
MW: 76.1 g/mol
Boiling Pt: 214 °C (760 mmHg)
Melting Pt: –32 °C
Density: 1.052 g/cm³
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00002949
CAS Number: 504-63-2
EINECS: 207-997-3
Merck Index: 12,09845

Order Now

Specification Test Results

Appearance Clear viscous liquid
Infrared spectrum Conforms
Refractive index 1.4380 to 1.4400 (20°C, 589 nm)
Water ≤0.2 % (K.F.)
GC ≥97.5 %
Color scale ≤20 APHA
UV at 400 nm A: ≤0.009
Residue after evaporation ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR