Propyl-4-hydroxybenzoate 99+%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Propyl 4-hydroxybenzoate, Propyl parahydroxybenzoate, Propylparaben, 4-Hydroxybenzoic acid propyl ester

131590050 131595000 131590025 131591000
ACRO131590050EA 41.2 EUR
ACRO131590050 ACRO131595000 ACRO131590025 ACRO131591000
Propyl-4-hydroxybenzoate 99+%
Propyl 4-hydroxybenzoate
Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 292
Melting Pt: 95…99 °C
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00002354
CAS Number: 94-13-3
EINECS: 202-307-7
Merck Index: 13,07958

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Melting point 95 to 99 °C
Sulfated ash ≤0.1 %
GC ≥99.0 %
Appearance of solution almost colorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR