1,1,2,2-Tetrachloroethane-[D₂] 99 atom % D NMR

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1,1,2,2-Tetrachloroethane-[²H₂]

133580050 133580200
ACRO133580050EA 111 EUR
ACRO133580050 ACRO133580200
1,1,2,2-Tetrachloroethane-[D₂] 99 atom % D NMR
1,1,2,2-Tetrachloroethane-[D2]
Formula: Cl₂CDCDCl₂
MW: 169.83 g/mol
Boiling Pt: 146
Melting Pt: –43 °C
Density: 1.61 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00037672
CAS Number: 33685-54-0
EINECS: 251-634-1
UN: 1702
ADR: 6.1 II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.03 %
D-Enrichment ≥98.5 %
NMR (19F-NMR) Consistent with structure
NMR (31P-NMR) Consistent with structure
Elemental Analysis 52.4 to 56.8 % C
Specific optical rotation +40° to +50° (20°C, 589 nm) (c=1 in methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR