DL-Tartaric acid 99.5%

Supplier: Thermo Fisher Scientific
Danger

137875000 137870025 137871000
ACRO137875000EA 139 EUR
ACRO137875000 ACRO137870025 ACRO137871000
DL-Tartaric acid 99.5%
DL-Tartaric acid

Appearance: White Crystalline powder

Formula: C₄H₆O₆
Melting Pt: 200…206 °C
Flash Pt: 210 °C
Storage Temperature: Ambient
MDL Number: MFCD00151254
CAS Number: 133-37-9

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 200 to 206 °C
Loss on drying ≤0.5 %
Sulfated ash ≤0.1 %
Specific optical rotation -0.5° to +0.5° (20 °C, 589 nm) (c=20, H2O)
Titration with NaOH ≥99.4 %
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR