1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone) 98%

Supplier: Thermo Fisher Scientific
Warning

138310250 138311000 138315000
ACRO138310250EA 34.5 EUR
ACRO138310250 ACRO138311000 ACRO138315000
1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone) 98%
1,2,3,4-Tetrahydronaphthalen-1-one (1-Tetralone, α-Tetralone)
Formula: C₁₀H₁₀O
Boiling Pt: 127 °C (13 mmHg)
Melting Pt: 5…6 °C
Density: 1,096 g/cm³
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00001688
CAS Number: 529-34-0
EINECS: 208-460-6

Order Now

Specification Test Results

Appearance Clear yellow to amber to brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5660 to 1.5710 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR