Benzo[b]thiophene 97%

Supplier: Thermo Fisher Scientific
Warning

138620050 138620250 138621000
ACRO138620050EA 41 EUR
ACRO138620050 ACRO138620250 ACRO138621000
Benzo[b]thiophene 97%
Benzo[b]thiophene
Formula: C₈H₆S
Boiling Pt: 221…222 °C (760 mmHg)
Melting Pt: 28…32 °C
Density: 1,14 g/cm³
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005864
CAS Number: 95-15-8
EINECS: 202-395-7
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to yellow to pink to orange Crystalline solid
Infrared spectrum Conforms
Melting point 28 to 33 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR