(±)-2-Mercaptopropionic acid 95%

Supplier: Thermo Fisher Scientific
Danger

138791000 138795000 138790025
ACRO138791000EA 44.3 EUR
ACRO138791000 ACRO138795000 ACRO138790025
(±)-2-Mercaptopropionic acid 95%
(±)-2-Mercaptopropionic acid
Formula: C₃H₆O₂S
Boiling Pt: 102 °C (16 mmHg)
Melting Pt: 10…14 °C
Density: 1,19 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00004862
CAS Number: 79-42-5
EINECS: 201-206-5
UN: 2936
ADR: 6.1,II
Merck Index: 12,09476

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4804 to 1.4824 (20°C, 589 nm)
Titration Iodimetric ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR