Thiosalicylic acid 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Thiobenzoic acid, 2-Mercaptobenzoic acid

138891000 138895000
ACRO138891000EA 97.3 EUR
ACRO138891000 ACRO138895000
Thiosalicylic acid 98%
Thiosalicylic acid
Formula: C₇H₆O₂S
MW: 154.19 g/mol
Boiling Pt: 230 °C (0.02 mmHg)
Melting Pt: 164…169 °C
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00004836
CAS Number: 147-93-3
EINECS: 205-704-3
ADR: 9
Merck Index: 12,09498

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 164 to 169 °C
Titration Argentometric 97.5 to 102.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR