Tri-O-acetyl-D-glucal 99%

Supplier: Thermo Fisher Scientific

139240250 139241000
ACRO139240250EA 97.9 EUR
ACRO139240250 ACRO139241000
Tri-O-acetyl-D-glucal 99%
Tri-O-acetyl-D-glucal

Appearance: White to off-white Crystalline powder

Formula: C₁₂H₁₆O₇
Melting Pt: 51…53 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063253
CAS Number: 2873-29-2

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (30°C, 15 h) (vacuum)
Sulfated ash ≤0.1 %
GC ≥98.5 %
Specific optical rotation -14.5° to -15.5° (20°C, 589 nm) (c=2, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR