Trifluoroacetic acid 99%, extra pure

Supplier: Thermo Fisher Scientific
Danger

139720010 139720250 139721000 139725000 139720025 139720100
ACRO139720010EA 469 EUR
ACRO139720010 ACRO139720250 ACRO139721000 ACRO139725000 ACRO139720025 ACRO139720100
Trifluoroacetic acid 99%, extra pure
Trifluoroacetic acid
Formula: C₂HF₃O₂
Boiling Pt: 72 °C (760 mmHg)
Melting Pt: –15 °C
Density: 1,535 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I
Merck Index: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.3 % (Coulometric)
Color scale ≤20 APHA
Titration with NaOH 99.0 to 101.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR