Trityl chloride 98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Trt-Cl, Chlorotriphenylmethane, Triphenylmethyl chloride, (Chloromethanetriyl)tribenzene, Triphenylchloromethane

140401000 140405000 140400025
ACRO140401000EA 65.5 EUR
ACRO140401000 ACRO140405000 ACRO140400025
Trityl chloride 98%
Trityl chloride
Formula: C₁₉H₁₅Cl
MW: 278.78 g/mol
Boiling Pt: 230…235 °C (20 mmHg)
Melting Pt: 110…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000813
CAS Number: 76-83-5
EINECS: 200-986-4
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Appearance White to off-white to yellow to light beige Powder or crystals
Infrared spectrum Conforms
Melting point 109 to 113 °C
Titration Argentometric ≥97.5 % (ex Cl)
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR