Tropinone 99%

Supplier: Thermo Fisher Scientific
Danger

140570100 140570050 140570500
ACRO140570100EA 83.2 EUR
ACRO140570100 ACRO140570050 ACRO140570500
Tropinone 99%
Tropinone
Formula: C₈H₁₃NO
Boiling Pt: 113 °C (25 mmHg)
Melting Pt: 41…44 °C
Flash Pt: 90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005549
CAS Number: 532-24-1
EINECS: 208-530-6
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Appearance Light yellow to brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 41 to 44 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR