Triphenylmethane 99+%

Supplier: Thermo Fisher Scientific

146380250 146381000 146385000
ACRO146380250EA 33.7 EUR
ACRO146380250 ACRO146381000 ACRO146385000
Triphenylmethane 99+%
Triphenylmethane
Formula: C₁₉H₁₆
Boiling Pt: 358…359 °C (754 mmHg)
Melting Pt: 90…94 °C
Density: 1,01 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00004763
CAS Number: 519-73-3
EINECS: 208-275-0
Merck Index: 12,09871

Order Now

Specification Test Results

Appearance Light yellow Powder
Infrared spectrum Conforms
Melting point 90 to 94 °C
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR