1,3-Diphenylisobenzofuran 97%

Supplier: Thermo Fisher Scientific

147580010 147580050
ACRO147580010EA 41.3 EUR
ACRO147580010 ACRO147580050
1,3-Diphenylisobenzofuran 97%
1,3-Diphenylisobenzofuran

Appearance: Yellow to green Crystalline solid or needles

Formula: C₂₀H₁₄O
Melting Pt: 129…133 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005931
CAS Number: 5471-63-6

Order Now

Specification Test Results

Appearance Yellow to green Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 128 to 137 °C
GC ≥96.0 %
pH 3.6 to 4.0 (0.5 % in water)
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR