2-Aminobenzothiazole 97%

Supplier: Thermo Fisher Scientific

148810050 148811000 148815000
ACRO148810050EA 30.5 EUR
ACRO148810050 ACRO148811000 ACRO148815000
2-Aminobenzothiazole 97%
2-Aminobenzothiazole
Formula: C₇H₆N₂S
Boiling Pt: 190…195 °C (0,05 mmHg)
Melting Pt: 129…134 °C
MDL Number: MFCD00005785
CAS Number: 136-95-8
EINECS: 205-268-4
Merck Index: 13,00423

Order Now

Specification Test Results

Appearance Beige to grey Powder or flakes
Infrared spectrum Conforms
Melting point 129 to 134 °C
Water ≤1 %
HPLC ≥97.0 %
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR