4'-Hydroxyacetophenone 98%

Supplier: Thermo Fisher Scientific
Warning

150400050 150401000 150405000
ACRO150400050EA 34.4 EUR
ACRO150400050 ACRO150401000 ACRO150405000
4'-Hydroxyacetophenone 98%
4'-Hydroxyacetophenone
Formula: C₈H₈O₂
Boiling Pt: 147…148 °C (3 mmHg)
Melting Pt: 107…111 °C
Density: 1,139 g/cm³ (25 °C)
Flash Pt: 166 °C
Storage Temperature: Ambient
MDL Number: MFCD00002359
CAS Number: 99-93-4
EINECS: 202-802-8

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 107 to 111 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR