4-Nitro-o-anisidine 99%

Supplier: Thermo Fisher Scientific
Warning

150640050 150641000 150645000
ACRO150640050EA 34.3 EUR
ACRO150640050 ACRO150641000 ACRO150645000
4-Nitro-o-anisidine 99%
4-Nitro-o-anisidine
Formula: C₇H₈N₂O₃
Boiling Pt: 364 °C (1013 hPa)
Melting Pt: 139…142 °C
Density: 1,2112 g/cm³ (156 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007363
CAS Number: 97-52-9
EINECS: 202-588-6

Order Now

Specification Test Results

Appearance Yellow to orange Crystalline powder
Infrared spectrum Conforms
Melting point 139 to 142 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR