Formaldehyde sodium bisulphite 95%

Supplier: Thermo Fisher Scientific

150690010 150690050 150690051
ACRO150690010EA 94.3 EUR
ACRO150690010 ACRO150690050 ACRO150690051
Formaldehyde sodium bisulphite 95%
Formaldehyde sodium bisulphite

Appearance: White to off-white Powder

Formula: CH₄O₄S
Melting Pt: 200 °C
Flash Pt: 184 °C
Storage Temperature: Ambient
MDL Number: MFCD00044664
CAS Number: 870-72-4

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration ≥94 %
HPLC ≥97.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR