2-tert-Butylhydroquinone 97%

Supplier: Thermo Fisher Scientific
Warning

150820020 150820050 150822500 150825000
ACRO150820020EA 414 EUR
ACRO150820020 ACRO150820050 ACRO150822500 ACRO150825000
2-tert-Butylhydroquinone 97%
2-tert-Butylhydroquinone
Formula: C₁₀H₁₄O₂
Boiling Pt: 273 °C (1013 hPa)
Melting Pt: 125…130 °C
Density: 1,05 g/cm³ (20 °C)
Flash Pt: 171 °C
Storage Temperature: Ambient
MDL Number: MFCD00002344
CAS Number: 1948-33-0
EINECS: 217-752-2
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to tan Crystalline powder (May contain black particles)
Infrared spectrum Conforms
Melting point 125 to 130 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR