Methyl red sodium salt, pure Indicator

Supplier: Thermo Fisher Scientific

Synonyms: Methyl red water soluble, 2-(4-Dimethylaminophenylazo)benzoic acid sodium salt, Sodium 2-(p-(dimethylamino)phenylazo)benzoate

151411000 151410250 151415000
ACRO151411000EA 82.5 EUR
ACRO151411000 ACRO151410250 ACRO151415000
Methyl red sodium salt, pure Indicator
Methyl red sodium salt
Formula: C₁₅H₁₄N₃NaO₂
MW: 291.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002426
CAS Number: 845-10-3
EINECS: 212-682-9
Merck Index: 13,06142

Order Now

Specification Test Results

Appearance Red to brown Powder
Infrared spectrum Conforms
Solubility (0.1% in water) clear to faint turbid solution
Visual transition interval from pH 4.2 (red) to pH 6.2 (yellow)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR