Phenol red sodium salt, water soluble, pure indicator

Supplier: Thermo Fisher Scientific
Warning

151440250 151440050 151441000
ACRO151440250EA 97 EUR
ACRO151440250 ACRO151440050 ACRO151441000
Phenol red sodium salt, water soluble, pure indicator
Phenol red sodium salt
Formula: C₁₉H₁₃NaO₅S
Melting Pt: 285 °C
Storage Temperature: Ambient
MDL Number: MFCD00066901
CAS Number: 34487-61-1
EINECS: 252-057-8
Merck Index: 12,07397

Order Now

Specification Test Results

Appearance Dark red to brown Powder
Infrared spectrum Conforms
Solubility (0.1 % in water) Clear
Specific extinction E (1%/1cm) ≥1600 (558 nm, in buffer)
Visual transition interval from pH 6.8 (yellow) to pH 8.2 (red)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR