4-Nitrophenethyl bromide 98%

Supplier: Thermo Fisher Scientific
Warning

151620050 151620250
ACRO151620050EA 59.8 EUR
ACRO151620050 ACRO151620250
4-Nitrophenethyl bromide 98%
4-Nitrophenethyl bromide
Formula: C₈H₈BrNO₂
Melting Pt: 67…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00007386
CAS Number: 5339-26-4
EINECS: 226-271-7

Order Now

Specification Test Results

Appearance Yellow Adhering crystalline powder or crystals
Infrared spectrum Conforms
Melting point 67 to 70 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR